Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007935.2 | NP_031961.1 | 10 | 1640 | ||
NM_027497.3 | NP_081773.1 | 10 | 1791 | |||
GenBank mRNA | AF079765.1 | - | 10 | 1619 | ||
AK040254.1 | - | 10 | 1791 | |||
AK148194.1 | - | 10 | 1609 | |||
AK149317.1 | - | 7 | 991 | |||
BC138622.1 | - | 10 | 1619 | 8861642 | ||
BC138623.1 | - | 10 | 1619 | 8861643 |