Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007936.3 | NP_031962.2 | 12 | 2146 | ||
GenBank mRNA | AK028509.1 | - | 12 | 2139 | ||
AK147698.1 | - | 12 | 2142 | |||
BC004782.1 | - | 12 | 2061 | 3588285 | ||
BC052164.1 | - | 12 | 2118 | 6512978 | ||
S57168.1 | - | 12 | 2146 | |||
X65138.1 | - | 12 | 2146 |