Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007937.3 | NP_031963.2 | 12 | 2750 | ||
GenBank mRNA | BC057401.1 | - | 11 | 2659 | 5697758 | |
U07357.1 | - | 12 | 2290 |