Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007939.2 | NP_031965.2 | 8 | 1708 | ||
GenBank mRNA | AK220393.1 | - | 6 | 1192 | ||
U72207.1 | - | 8 | 1717 |