Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290753.1 | NP_001277682.1 | 3 | 650 | ||
NM_010142.3 | NP_034272.1 | 3 | 650 | |||
GenBank mRNA | AK017630.1 | - | 3 | 614 | ||
BC043088.1 | - | 3 | 574 | 6418088 | ||
BC062924.1 | - | 3 | 595 | |||
L25890.1 | - | 4 | 492 |