Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010143.1 | NP_034273.1 | 3 | 761 | ||
GenBank mRNA | AK144414.1 | - | 3 | 777 | ||
AK158043.1 | - | 3 | 1822 | |||
BC014822.1 | - | 3 | 762 | 3673003 | ||
BC053085.1 | - | 3 | 761 | 5708827 | ||
U11493.1 | - | 1 | 221 | |||
Z49086.1 | - | 3 | 619 |