Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146351.1 | NP_001139823.1 | 15, 16 | 3103 | ||
NM_007680.4 | NP_031706.3 | 15, 16 | 3099 | |||
GenBank mRNA | AK076442.1 | - | 15, 16 | 2926 | ||
AK087423.1 | - | 15, 16 | 3014 | |||
AK163823.1 | - | 6, 7 | 821 | |||
BC031924.1 | - | 14, 15 | 2711 | 4911959 | ||
L77867.1 | - | 15, 16 | 2793 |