Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000189.4 | NP_000180.2 | 10 | 3403 | ||
XM_005264280.1 | XP_005264337.1 | 10 | 1883 | |||
XM_011532807.1 | XP_011531109.1 | 9 | 1787 | |||
XM_017003945.1 | XP_016859434.1 | 9 | 1787 | |||
GenBank mRNA | AF148513.1 | - | 10 | 1926 | ||
AK290357.1 | - | 10 | 1906 | |||
BC021116.1 | - | 10 | 1921 | 4652321 | ||
BC064369.1 | - | 10 | 1919 | 5586773 | ||
CR749202.1 | - | 10 | 1583 |