Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159964.1 | NP_001153436.1 | 2 | 333 | ||
NM_007943.3 | NP_031969.1 | 14 | 1282 | |||
GenBank mRNA | AK028361.1 | - | 136 | |||
AK083176.1 | - | 2 | 317 | |||
AY190610.1 | - | 14 | 1255 | |||
BC048783.1 | - | 14 | 1256 | 6466448 | ||
CF533628.1 | - | 2 | 333 | |||
L21768.1 | - | 14 | 1255 |