Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003817.1 | NP_001003817.1 | 18 | 2318 | ||
GenBank mRNA | AK031099.1 | - | 18 | 2186 | ||
AK031542.1 | - | 10 | 1016 | |||
AK083669.1 | - | 17 | 2268 | |||
AK129487.1 | - | 18 | 2288 | |||
AK142296.1 | - | 18 | 2312 | |||
BC046553.1 | - | 4 | 673 | |||
BC046811.1 | - | 17 | 2390 | 5702040 | ||
BC053078.1 | - | 17 | 2390 | 5702040 |