Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003817.1 | NP_001003817.1 | 11, 12 | 1498 | ||
GenBank mRNA | AK031099.1 | - | 11, 12 | 1366 | ||
AK031542.1 | - | 3, 4 | 196 | |||
AK083669.1 | - | 11, 12 | 1366 | |||
AK129487.1 | - | 11, 12 | 1468 | |||
AK142296.1 | - | 11, 12 | 1490 | |||
BC046811.1 | - | 11, 12 | 1489 | 5702040 | ||
BC053078.1 | - | 11, 12 | 1489 | 5702040 |