Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003817.1 | NP_001003817.1 | 22 | 2858 | ||
GenBank mRNA | AK031099.1 | - | 22 | 2726 | ||
AK031542.1 | - | 14 | 1556 | |||
AK083669.1 | - | 21 | 2808 | |||
AK129487.1 | - | 22 | 2828 | |||
AK142296.1 | - | 22 | 2852 | |||
BC027080.1 | - | 4 | 470 | |||
BC046553.1 | - | 8 | 1213 | |||
BC046811.1 | - | 21 | 2930 | 5702040 | ||
BC053078.1 | - | 21 | 2930 | 5702040 | ||
U71126.1 | - | 3 | 324 |