Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ereg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007950.2 | NP_031976.1 | 2, 3 | 277 | ||
GenBank mRNA | BB612312.1 | - | 2, 3 | 272 | ||
BC027838.1 | - | 2, 3 | 253 | 5325124 | ||
CJ056327.1 | - | 2, 3 | 278 | |||
D30782.1 | - | 2, 3 | 272 |