Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278647.1 | NP_001265576.1 | 3 | 906 | ||
NM_001278648.1 | NP_001265577.1 | 3 | 927 | |||
NM_005341.3 | NP_005332.1 | 3 | 993 | |||
XM_017001110.1 | XP_016856599.1 | 3 | 978 | |||
XM_017001111.1 | XP_016856600.1 | 3 | 978 | |||
XM_017001112.1 | XP_016856601.1 | 3 | 983 | |||
XM_017001113.1 | XP_016856602.1 | 3 | 924 | |||
XM_017001114.1 | XP_016856603.1 | 3 | 911 | |||
GenBank mRNA | AK290156.1 | - | 3 | 901 | ||
AK312786.1 | - | 3 | 909 | |||
BC013573.2 | - | 2 | 852 | 3906038 | ||
L16896.1 | - | 3 | 933 |