Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB560752.1 | - | 5 | 1012 | ||
AB560753.1 | - | 4 | 875 | |||
AB560754.1 | - | 4 | 875 | |||
AB560755.1 | - | 4 | 875 | |||
AB560756.1 | - | 4 | 875 | |||
AK036627.1 | - | 5 | 986 | |||
AK039911.1 | - | 8 | 1489 | |||
AK041525.1 | - | 6 | 1134 | |||
AK054182.1 | - | 5 | 1017 | |||
AK077236.1 | - | 5 | 991 | |||
AK087638.1 | - | 5 | 966 | |||
AK139745.1 | - | 6 | 1546 | |||
EU791538.1 | - | 5 | 930 | |||
EU791540.1 | - | 5 | 1055 | |||
M38651.1 | - | 5 | 994 |