Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016707.3 | NP_057916.1 | 4, 5 | 2563 | ||
GenBank mRNA | AF051525.1 | - | 4, 5 | 2501 | ||
AF169036.1 | - | 2, 3 | 1704 | |||
BC010585.1 | - | 1, 2 | 1577 | 2631265 |