Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163394.1 | NP_001156866.1 | 2 | 601 | ||
NM_001163395.1 | NP_001156867.1 | 2 | 353 | |||
NM_001163396.2 | NP_001156868.1 | 3 | 375 | |||
NM_007965.3 | NP_031991.3 | 2 | 601 | |||
GenBank mRNA | AF279662.1 | - | 1 | 112 | ||
AK134563.1 | - | 2 | 353 | |||
AK137823.1 | - | 2 | 190 | |||
AK151633.1 | - | 3 | 376 | |||
BC059810.1 | - | 2 | 601 | 6828523 | ||
BC096017.1 | - | 2 | 494 | |||
U72519.1 | - | 2 | 364 |