Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ewsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283061.1 | NP_001269990.1 | 16 | 1942 | ||
NM_001283062.1 | NP_001269991.1 | 14 | 1828 | |||
NM_007968.3 | NP_031994.2 | 16 | 1939 | |||
GenBank mRNA | AK014366.1 | - | 8 | 908 | ||
AK019460.1 | - | 16 | 1888 | |||
AK034755.1 | - | 16 | 1915 | |||
AK049743.1 | - | 14 | 1807 | |||
AK147103.1 | - | 16 | 1906 | |||
AK150345.1 | - | 16 | 1900 | |||
AK151625.1 | - | 16 | 1910 | |||
AK153276.1 | - | 16 | 1900 | |||
BC068226.1 | - | 16 | 1909 | 5707017 | ||
CB205713.1 | - | 2 | 273 | |||
X79233.1 | - | 16 | 1928 |