Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ewsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283061.1 | NP_001269990.1 | 7 | 744 | ||
NM_001283062.1 | NP_001269991.1 | 7 | 744 | |||
NM_001283063.1 | NP_001269992.1 | 7 | 744 | |||
NM_007968.3 | NP_031994.2 | 7 | 744 | |||
GenBank mRNA | AK019460.1 | - | 7 | 693 | ||
AK034755.1 | - | 7 | 720 | |||
AK049743.1 | - | 7 | 724 | |||
AK146791.1 | - | 7 | 720 | |||
AK147103.1 | - | 7 | 711 | |||
AK147909.1 | - | 7 | 701 | |||
AK150345.1 | - | 7 | 702 | |||
AK151625.1 | - | 7 | 715 | |||
AK153276.1 | - | 7 | 702 | |||
BC068226.1 | - | 7 | 711 | 5707017 | ||
X79233.1 | - | 7 | 733 |