Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007969.4 | NP_031995.3 | 3 | 435 | ||
GenBank mRNA | AK145032.1 | - | 3 | 331 | ||
BC010986.1 | - | 3 | 435 | 4038377 | ||
BE310436.1 | - | 3 | 296 | |||
EH099462.1 | - | 3 | 462 | |||
EH105860.1 | - | 3 | 462 | |||
EH109450.1 | - | 3 | 341 | |||
X93037.1 | - | 3 | 333 |