Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ext1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010162.2 | NP_034292.2 | 1 | 1166 | ||
GenBank mRNA | AK132327.1 | - | 1 | 1143 | ||
BC004741.1 | - | 1 | 530 | 3499743 | ||
U78539.1 | - | 1 | 670 | |||
X96639.1 | - | 1 | 940 |