Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ext2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010163.3 | NP_034293.2 | 2 | 284 | ||
GenBank mRNA | AK082572.1 | - | 2 | 266 | ||
AK084772.1 | - | 2 | 266 | |||
AK148090.1 | - | 2 | 273 | |||
AK148369.1 | - | 1 | 45 | |||
AK150155.1 | - | 2 | 276 | |||
AK152265.1 | - | 384 | ||||
AK152840.1 | - | 384 | ||||
AK164342.1 | - | 2 | 270 | |||
BC006597.1 | - | 2 | 257 | 3965079 | ||
U67837.1 | - | 2 | 259 | |||
U72141.1 | - | 2 | 263 |