Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eya2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271962.1 | NP_001258891.1 | 4 | 308 | ||
NM_001271963.1 | NP_001258892.1 | 4 | 484 | |||
NM_010165.3 | NP_034295.1 | 4 | 485 | |||
GenBank mRNA | AK137718.1 | - | 4 | 480 | ||
AK141765.1 | - | 4 | 484 | |||
BC003755.1 | - | 4 | 485 | 3497245 | ||
CF724561.1 | - | 3 | 184 | |||
U61111.1 | - | 3 | 226 | |||
U71208.1 | - | 4 | 310 | |||
U81603.1 | - | 3 | 225 |