Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010171.3 | NP_034301.3 | 4 | 729 | ||
GenBank mRNA | AK137642.1 | - | 4 | 658 | ||
AK144872.1 | - | 4 | 658 | |||
AY500273.1 | - | 1 | 147 | |||
BC016397.1 | - | 4 | 720 | 3584897 | ||
BC024886.1 | - | 4 | 729 | 3591823 | ||
CT010367.1 | - | 4 | 551 | |||
M26071.1 | - | 4 | 685 | |||
M57896.1 | - | 4 | 585 |