Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007978.3 | NP_032004.2 | 1 | 1308 | ||
GenBank mRNA | AF299331.1 | - | 1 | 1337 | ||
AK033184.1 | - | 1 | 1311 | |||
AK046025.1 | - | 1 | 1324 | |||
AK046459.1 | - | 1 | 1323 | |||
AK054140.1 | - | 1 | 1309 | |||
AK166334.1 | - | 1 | 1324 | |||
BC131938.1 | - | 1 | 1336 | 40130215 | ||
BC131940.1 | - | 1 | 1336 | 40130217 | ||
M83118.1 | - | 1 | 1590 |