Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK149372.1 | - | 7, 8 | 867 | ||
BC125617.1 | - | 7, 8 | 865 | |||
BC132393.1 | - | 7, 8 | 865 | |||
M23109.1 | - | 7, 8 | 830 | |||
M26236.1 | - | 2, 3 | 329 |