Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Faah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010173.4 | NP_034303.3 | 12, 13 | 1597 | ||
GenBank mRNA | AK004985.1 | - | 11, 12 | 2121 | ||
AK079598.1 | - | 12, 13 | 1598 | |||
AK080603.1 | - | 12, 13 | 1430 | |||
AK138481.1 | - | 12, 13 | 1217 | |||
BC006863.1 | - | 12, 13 | 1376 | 3595191 | ||
BC052321.1 | - | 12, 13 | 1639 | 6515634 | ||
U82536.1 | - | 12, 13 | 1358 |