Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010176.4 | NP_034306.2 | 5 | 640 | ||
GenBank mRNA | AK143759.1 | - | 5 | 469 | ||
AK158808.1 | - | 5 | 526 | |||
BC010767.1 | - | 5 | 425 | 4242421 | ||
M84145.1 | - | 5 | 466 | |||
Z11774.1 | - | 5 | 452 |