Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fscn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007984.2 | NP_032010.2 | 3 | 1100 | ||
GenBank mRNA | AK083935.1 | - | 3 | 1101 | ||
AK167670.1 | - | 3 | 1100 | |||
BC010338.1 | - | 3 | 1086 | |||
BC029281.1 | - | 3 | 1088 | |||
BC037137.1 | - | 3 | 1097 | |||
BC052408.1 | - | 3 | 1089 | 5716179 | ||
L33726.1 | - | 3 | 1084 |