Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042673.2 | NP_001036138.1 | 4 | 633 | ||
NM_001282942.1 | NP_001269871.1 | 3 | 436 | |||
NM_007985.3 | NP_032011.2 | 4 | 633 | |||
GenBank mRNA | AK035488.1 | - | 4 | 387 | ||
AK042226.1 | - | 4 | 603 | |||
AK080697.1 | - | 614 | ||||
AK082717.1 | - | 3 | 414 | |||
AK147025.1 | - | 4 | 613 | |||
AK163161.1 | - | 4 | 634 | |||
AK196042.1 | - | 4 | 363 | |||
BC038609.1 | - | 3 | 418 | 5375091 | ||
L08266.1 | - | 4 | 614 |