Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080387.2 | NP_001073856.1 | 2 | 268 | ||
NM_001284195.1 | NP_001271124.1 | 2 | 268 | |||
NM_001284196.1 | NP_001271125.1 | 2 | 268 | |||
NM_010178.3 | NP_034308.1 | 2 | 268 | |||
GenBank mRNA | AB015894.1 | - | 2 | 179 | ||
AB015895.1 | - | 2 | 176 | |||
AF042383.1 | - | 2 | 180 | |||
AF060490.1 | - | 2 | 203 | |||
AK005295.1 | - | 2 | 210 | |||
AK014345.1 | - | 2 | 207 | |||
AK134076.1 | - | 2 | 238 | |||
AK134289.1 | - | 2 | 238 | |||
AK135316.1 | - | 2 | 213 | |||
AK149716.1 | - | 2 | 195 | |||
AK150072.1 | - | 2 | 195 | |||
AK150734.1 | - | 2 | 202 | |||
AK151567.1 | - | 2 | 207 | |||
AK161097.1 | - | 2 | 227 | |||
AK168524.1 | - | 2 | 226 | |||
AK168558.1 | - | 2 | 226 | |||
AK168571.1 | - | 2 | 191 | |||
AK169071.1 | - | 2 | 195 | |||
BC006670.1 | - | 2 | 220 | |||
BC037591.1 | - | 2 | 189 | 5004611 | ||
BC043060.1 | - | 2 | 592 | 6419883 | ||
BC083082.1 | - | 2 | 180 | 6511807 | ||
BY717856.1 | - | 2 | 238 | |||
CF914989.1 | - | 2 | 184 |