Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010180.2 | NP_034310.2 | 8 | 1019 | ||
GenBank mRNA | AK035388.1 | - | 8 | 972 | ||
AK083573.1 | - | 8 | 1022 | |||
AK085989.1 | - | 8 | 962 | |||
AK086451.1 | - | 8 | 1020 | |||
AK137511.1 | - | 8 | 962 | |||
AK159646.1 | - | 8 | 974 | |||
BC007140.1 | - | 8 | 963 | 3495754 | ||
EU543224.1 | - | 8 | 957 | |||
X70854.1 | - | 8 | 1016 |