Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081437.1 | NP_001074906.1 | 17 | 3356 | ||
NM_007992.2 | NP_032018.2 | 18 | 3497 | |||
GenBank mRNA | AK087922.1 | - | 6 | 740 | ||
AK159361.1 | - | 17 | 3668 | |||
AK161522.1 | - | 17 | 3299 | |||
AK168688.1 | - | 17 | 3294 | |||
BC005443.1 | - | 17 | 3336 | 3490759 | ||
X75285.1 | - | 18 | 3397 |