Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242524.1 | NP_001229453.1 | 3 | 481 | ||
NM_001242525.1 | NP_001229454.1 | 3 | 405 | |||
NM_033554.3 | NP_291032.2 | 2 | 390 | |||
GenBank mRNA | AB209058.1 | - | 2 | 295 | ||
AK292709.1 | - | 2 | 297 | |||
BC009956.2 | - | 2 | 314 | 4309471 | ||
BI553587.1 | - | 3 | 410 | |||
BP338039.1 | - | 2 | 390 | |||
CD516536.1 | - | 3 | 413 | |||
M27487.1 | - | 2 | 318 | |||
X00457.1 | - | 2 | 203 |