Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010185.4 | NP_034315.1 | 2 | 186 | ||
GenBank mRNA | AK155600.1 | - | 2 | 143 | ||
BC034163.1 | - | 126 | 5065647 | |||
BY173505.1 | - | 2 | 186 | |||
CK791068.1 | - | 2 | 136 | |||
J05020.1 | - | 2 | 124 |