Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253737.1 | NP_001240666.1 | 10 | 780 | ||
NM_001253739.1 | NP_001240668.1 | 10 | 791 | |||
NM_001253743.1 | NP_001240672.1 | 11 | 840 | |||
NM_001253745.1 | NP_001240674.1 | 9 | 717 | |||
NM_001253746.1 | NP_001240675.1 | 9 | 717 | |||
NM_001253747.1 | NP_001240676.1 | 10 | 801 | |||
NM_013517.4 | NP_038545.1 | 11 | 921 | |||
GenBank mRNA | AK156654.1 | - | 11 | 924 | ||
AK172591.1 | - | 11 | 924 | |||
AY069980.1 | - | 9 | 683 | |||
AY069981.1 | - | 9 | 683 | |||
BC119164.1 | - | 10 | 788 | 8733913 | ||
BC119190.1 | - | 10 | 788 | 8733939 | ||
M34163.1 | - | 11 | 923 | |||
M99371.1 | - | 11 | 913 | |||
S67072.1 | - | 10 | 772 | |||
S67073.1 | - | 9 | 652 | |||
S67074.1 | - | 10 | 691 | |||
X64223.1 | - | 10 | 780 | |||
X64224.1 | - | 10 | 791 |