Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253737.1 | NP_001240666.1 | 3 | 187 | ||
NM_001253739.1 | NP_001240668.1 | 3 | 198 | |||
NM_001253743.1 | NP_001240672.1 | 4 | 247 | |||
NM_001253745.1 | NP_001240674.1 | 3 | 187 | |||
NM_001253746.1 | NP_001240675.1 | 3 | 187 | |||
NM_001253747.1 | NP_001240676.1 | 3 | 208 | |||
NM_013517.4 | NP_038545.1 | 4 | 328 | |||
GenBank mRNA | AK080159.1 | - | 3 | 520 | ||
AK156654.1 | - | 4 | 331 | |||
AK172591.1 | - | 4 | 331 | |||
AY069980.1 | - | 3 | 153 | |||
AY069981.1 | - | 3 | 153 | |||
BC119164.1 | - | 3 | 195 | 8733913 | ||
BC119190.1 | - | 3 | 195 | 8733939 | ||
M34163.1 | - | 4 | 330 | |||
M99371.1 | - | 4 | 320 | |||
S67072.1 | - | 3 | 179 | |||
S67073.1 | - | 2 | 59 | |||
S67074.1 | - | 3 | 98 | |||
X64223.1 | - | 3 | 187 | |||
X64224.1 | - | 3 | 198 |