Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fem1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010192.4 | NP_034322.3 | 1 | 1969 | ||
GenBank mRNA | AK005041.1 | - | 1 | 1928 | ||
AK009354.1 | - | 1 | 262 | |||
AK034412.1 | - | 1 | 1944 | |||
AK075860.1 | - | 1 | 93 | |||
AK080978.1 | - | 1 | 1941 | |||
AK129000.1 | - | 1 | 1952 | |||
AK147879.1 | - | 1 | 1954 | |||
AK155363.1 | - | 1 | 1953 | |||
AK182414.1 | - | 1 | 119 | |||
AK183479.1 | - | 1 | 147 | |||
AK190230.1 | - | 1 | 118 | |||
BC009161.1 | - | 1 | 1936 | 2811079 | ||
BC054382.1 | - | 1 | 1940 | 6469158 | ||
BC110669.1 | - | 1 | 1923 | 5042239 |