Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fer | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037997.3 | NP_001033086.2 | 11 | 1650 | ||
NM_001286415.1 | NP_001273344.1 | 10 | 1478 | |||
NM_008000.2 | NP_032026.2 | 6 | 932 | |||
GenBank mRNA | AK082799.1 | - | 8 | 1143 | ||
AK157828.1 | - | 11 | 1614 | |||
BC051249.1 | - | 3 | 394 | |||
BC058100.1 | - | 10 | 1478 | 6813507 | ||
M32054.1 | - | 6 | 932 | |||
U76762.1 | - | 10 | 1583 |