Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010194.2 | NP_034324.2 | 6 | 895 | ||
GenBank mRNA | AK004587.1 | - | 6 | 887 | ||
AK143639.1 | - | 6 | 845 | |||
AK155861.1 | - | 6 | 842 | |||
AK170418.1 | - | 6 | 836 | |||
BC100306.1 | - | 5 | 993 | |||
BC129919.1 | - | 5 | 831 | 40111855 | ||
X12616.1 | - | 5 | 835 |