Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008001.4 | NP_032027.2 | 10 | 2335 | ||
GenBank mRNA | AK148151.1 | - | 10 | 2336 | ||
AK219664.1 | - | 1 | 136 | |||
BC004838.1 | - | 2 | 225 | |||
BC011462.1 | - | 10 | 2272 | 3674293 | ||
U22325.1 | - | 10 | 1934 |