Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291104.1 | NP_001278033.1 | 1 | 196 | ||
NM_010198.2 | NP_034328.1 | 1 | 196 | |||
GenBank mRNA | BC066859.1 | - | 1 | 196 | 30101881 | |
U66203.1 | - | 1 | 105 |