Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DQA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020056.4 | NP_064440.1 | 2 | 275 | ||
GenBank mRNA | M95727.1 | - | 1 | 115 | ||
M95728.1 | - | 1 | 115 |