Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008007.2 | NP_032033.2 | 3 | 468 | ||
GenBank mRNA | AK021062.1 | - | 2 | 97 | ||
BC117059.1 | - | 3 | 426 | 40126378 | ||
BC117061.1 | - | 3 | 426 | 40126380 | ||
BU923457.1 | - | 3 | 468 |