Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010208.4 | NP_034338.3 | 4 | 460 | ||
GenBank mRNA | AK036438.1 | - | 4 | 382 | ||
AK036476.1 | - | 4 | 632 | |||
AK038141.1 | - | 4 | 366 | |||
AK089812.1 | - | 4 | 430 | |||
AK155902.1 | - | 4 | 436 | |||
AK157640.1 | - | 414 | ||||
AK171824.1 | - | 4 | 414 | |||
BC057863.1 | - | 4 | 329 | 5317449 | ||
X16440.1 | - | 4 | 395 | |||
X52191.1 | - | 4 | 367 |