Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA797426.1 | - | 1, 2 | 104 | ||
AF047699.1 | - | 4, 5 | 599 | |||
AF055573.1 | - | 465 | ||||
BC012662.1 | - | 2, 3 | 324 | 4036815 | ||
CJ053929.1 | - | 358 |