Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301412.1 | NP_001288341.1 | 9 | 1170 | ||
NM_008017.4 | NP_032043.3 | 9 | 1094 | |||
GenBank mRNA | AJ534939.1 | - | 8 | 925 | ||
AK017305.1 | - | 9 | 1139 | |||
AK017747.1 | - | 9 | 1072 | |||
AK028233.1 | - | 9 | 1136 | |||
AK145341.1 | - | 9 | 1124 | |||
AK152090.1 | - | 9 | 1122 | |||
BC011064.1 | - | 9 | 1032 | |||
BC021847.1 | - | 9 | 1184 | |||
BC094380.1 | - | 9 | 1066 | 30543190 |