Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010217.2 | NP_034347.2 | 5 | 1233 | ||
GenBank mRNA | BC006783.1 | - | 5 | 1154 | 3589136 | |
BQ892143.1 | - | 2 | 370 | |||
M70642.1 | - | 5 | 1086 | |||
M80263.1 | - | 5 | 1152 |