Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK297032.1 | - | 3 | 418 | ||
AK304429.1 | - | 3 | 301 | |||
AK313123.1 | - | 3 | 410 | |||
BC032350.1 | - | 3 | 386 | 5214156 | ||
BC071659.1 | - | 3 | 394 | 5162925 | ||
BM849755.1 | - | 3 | 456 | |||
CR457013.1 | - | 3 | 346 | |||
J00194.1 | - | 3 | 372 | |||
K01171.1 | - | 3 | 360 | |||
M60334.1 | - | 3 | 373 | |||
V00523.1 | - | 2 | 255 |
Gene Symbol: HLA-DQA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF522250.1 | - | 366 |