Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB241120.1 | - | 4 | 378 | ||
AB241121.1 | - | 4 | 271 | |||
AF483488.1 | - | 3 | 143 | |||
AF483489.1 | - | 3 | 143 | |||
AK002777.1 | - | 3 | 250 | |||
AK010693.1 | - | 3 | 274 | |||
AK019362.1 | - | 3 | 278 | |||
AK145400.1 | - | 3 | 267 | |||
AK145986.1 | - | 2 | 300 | |||
AK151047.1 | - | 1 | 61 | |||
AK154751.1 | - | 3 | 222 | |||
AK168333.1 | - | 3 | 258 | |||
AK169186.1 | - | 3 | 181 | |||
AK169242.1 | - | 3 | 265 | |||
BC004671.1 | - | 3 | 225 | 3498682 | ||
BY065108.1 | - | 3 | 352 | |||
CX239711.1 | - | 4 | 358 | |||
X60203.1 | - | 3 | 240 |